Learn More
Silicon(IV) oxide, 15% in H{2}O, colloidal dispersion
Description
High-temperature binders, anti-slip coatings for paper, petrochemical catalyst support and binder, polishing agent, abrasion resistant coatingsSilicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of food and pharmaceutical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Particles in Liquid |
| Quantity | 1 kg |
| Solubility Information | Immiscible with water. |
| IUPAC Name | dioxosilane |
| Formula Weight | 60.09 |
| Percent Purity | 15% |
| Chemical Name or Material | Silicon(IV) oxide, 15% in H2O |
Safety and Handling
| Hazard Category | H373: May cause damage to organs through prolonged or repeated exposure. |
| EINECSNumber | 231-545-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only