missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) nitride, alpha-phase, ca. 90%

CAS: 12033-89-5 | N4Si3 | 140.28 g/mol

Supplier:  Thermo Scientific Chemicals 04294930

 View more versions of this product

Catalog No. AA4294930


Only null left
Explore available promotions

Description

Description

Silicon(IV) nitride is used in the automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from this show good shock resistance. Similarly bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and in the manufacture of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silicon (IV) nitride
alpha-phase
Gray
250 g
MFCD00011230
silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
HQVNEWCFYHHQES-UHFFFAOYSA-N
Silicon nitride
3084099
95%
Odorless
1900°C (sublimation)
12033-89-5
Powder
N4Si3
14,8497
Insoluble in water.
N12[Si]34N5[Si]11N3[Si]25N41
140.28
140.28
Si3N4 85% min (alpha form)
3.2 g/mL

Chemical Identifiers

12033-89-5
140.28
HQVNEWCFYHHQES-UHFFFAOYSA-N
3084099
N12[Si]34N5[Si]11N3[Si]25N41
N4Si3
MFCD00011230
silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
Silicon nitride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-796-8

RTECSNumber : VW0650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only