missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) nitride, alpha-phase, ca. 90%

Catalog No. AA4294918
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4294918 50 g
AA4294930 250 g
AA42949A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4294918 Supplier Thermo Scientific Chemicals Supplier No. 04294918

CAS: 12033-89-5 | N4Si3 | 140.28 g/mol

Silicon(IV) nitride is used in the automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from this show good shock resistance. Similarly bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and in the manufacture of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12033-89-5
Molecular Formula N4Si3
Molecular Weight (g/mol) 140.28
MDL Number MFCD00011230
InChI Key HQVNEWCFYHHQES-UHFFFAOYSA-N
Synonym silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
PubChem CID 3084099
IUPAC Name Silicon nitride
SMILES N12[Si]34N5[Si]11N3[Si]25N41

Specifications

Melting Point 1900°C (sublimation)
Color Gray
Physical Form Powder
Quantity 50 g
Merck Index 14,8497
Solubility Information Insoluble in water.
Formula Weight 140.28
Percent Purity 95%
Concentration or Composition (by Analyte or Components) Si3N4 85% min (alpha form)
Odor Odorless
Density 3.2 g/mL
Chemical Name or Material Silicon (IV) nitride, alpha-phase
Show More Show Less
EINECSNumber 234-796-8
RTECSNumber VW0650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only