missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) chloride, 99.998% (metals basis)

Catalog No. AA40148AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40148AC 25 mL
AA40148AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40148AC Supplier Thermo Scientific Chemicals Supplier No. 040148AC

CAS: 10026-04-7 | Cl4Si | 169.89 g/mol

Silicon(IV) chloride is used as semiconductor grade.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10026-04-7
Molecular Formula Cl4Si
Molecular Weight (g/mol) 169.89
MDL Number MFCD00011229
InChI Key FDNAPBUWERUEDA-UHFFFAOYSA-N
Synonym silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4
PubChem CID 24816
IUPAC Name tetrachlorosilane
SMILES Cl[Si](Cl)(Cl)Cl

Specifications

Boiling Point 57.6°C
Melting Point -70°C
Color Colorless
Physical Form Liquid
Quantity 25 mL
Linear Formula SiCl4
UN Number UN1818
Merck Index 14,8500
Solubility Information Reacts with water. Miscible with benzene,ether,chloroform.
Formula Weight 169.9
Percent Purity 99.998%
Odor Acrid
Sensitivity Moisture sensitive
Density 1.483 g/mL
Assay (metals basis)
Chemical Name or Material Silicon(IV) chloride
Show More Show Less
Hazard Category H301+H331-H314-H335
Hazard Statement GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SILICON TETRACHLORIDE
EINECSNumber 233-054-0
RTECSNumber VW0525000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only