missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) chloride, 99.8+%

Catalog No. p-221875
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201165000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC201165000 Supplier Thermo Scientific Chemicals Supplier No. 201165000
Only null left

CAS: 10026-04-7 | Cl4Si | 169.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10026-04-7
Molecular Formula Cl4Si
Molecular Weight (g/mol) 169.89
MDL Number MFCD00011229
InChI Key FDNAPBUWERUEDA-UHFFFAOYSA-N
Synonym silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4
PubChem CID 24816
IUPAC Name tetrachlorosilane
SMILES Cl[Si](Cl)(Cl)Cl

Specifications

Boiling Point 58.0°C
Melting Point -70.0°C
Color Colorless
Physical Form Liquid
Quantity 500 mL
Assay Percent Range 99.8+%
Linear Formula SiCl4
Fieser 10,347; 11,466; 16,317
Merck Index 15, 8639
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with benzene, ether, chloroform,, petroleum ether, carbon tetrachloride,, tin tetrachloride, sulfur, mono- and dichlorides, decomposes by alcohol with evolution of hydrogen, chloride
Formula Weight 169.9
Percent Purity ≥99.8%
Packaging Glass bottle
Refractive Index 1.413
Specific Gravity 1.48
Density 1.4800g/mL
Chemical Name or Material Silicon(IV) chloride, 99.8+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
EINECSNumber 233-054-0
RTECSNumber VW0525000
TSCA TSCA
Recommended Storage Product may develop some turbidity or precipitate

RUO – Research Use Only