missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) acetate

CAS: 562-90-3 | C8H12O8Si | 264.26 g/mol

$271.86 - $1057.42

Chemical Identifiers

CAS 562-90-3
Molecular Formula C8H12O8Si
Molecular Weight (g/mol) 264.26
MDL Number MFCD00026184
InChI Key YZVRVDPMGYFCGL-UHFFFAOYSA-N
Synonym silicon tetraacetate, tetraacetoxysilane, tetraacetyl orthosilicate, unii-vz7lp47epp, vz7lp47epp, silicon iv acetate, anhydrous, acetic acid, 1,1',1,1'-tetraanhydride with silicic acid h4sio4, triacetoxysilyl acetate, silanetetrol, tetraacetate
PubChem CID 68419
IUPAC Name triacetyloxysilyl acetate
SMILES CC(=O)O[Si](OC(C)=O)(OC(C)=O)OC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1470318
View Documents
Thermo Scientific Chemicals
01470318
50 g
Each for $271.86
Only null left
 
AA1470330
View Documents
Thermo Scientific Chemicals
01470330
250 g
Each for $1,057.42
Only null left
 
Description

Description

Silicon tetraacetate is used in the preparation of silicon dioxide thin films by a direct photochemical vapor deposition method. It serves as a precursor to prepare silicon complexes with monofunctional bidentate Schiff bases. It is also used as an alternative to silicon hydride and alkoxide for low-temperature silicon dioxide production. Further, it reacts with ethanol in the absence of water to get silica gel and ethyl acetate. In addition, it is employed as a sol-gel precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

562-90-3
264.26
YZVRVDPMGYFCGL-UHFFFAOYSA-N
68419
CC(=O)O[Si](OC(C)=O)(OC(C)=O)OC(C)=O
C8H12O8Si
MFCD00026184
silicon tetraacetate, tetraacetoxysilane, tetraacetyl orthosilicate, unii-vz7lp47epp, vz7lp47epp, silicon iv acetate, anhydrous, acetic acid, 1,1',1,1'-tetraanhydride with silicic acid h4sio4, triacetoxysilyl acetate, silanetetrol, tetraacetate
triacetyloxysilyl acetate

Specifications

160°C to 170°C (decomposition)
562-90-3
50 g
MFCD00026184
14,8498
Soluble in acetone and benzene.
CC(=O)O[Si](OC(C)=O)(OC(C)=O)OC(C)=O
264.26
264.27
Silicon(IV) acetate
110°C
Crystalline
C8H12O8Si
UN3261
silicon tetraacetate, tetraacetoxysilane, tetraacetyl orthosilicate, unii-vz7lp47epp, vz7lp47epp, silicon iv acetate, anhydrous, acetic acid, 1,1',1,1'-tetraanhydride with silicic acid h4sio4, triacetoxysilyl acetate, silanetetrol, tetraacetate
YZVRVDPMGYFCGL-UHFFFAOYSA-N
triacetyloxysilyl acetate
68419
Moisture sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 209-239-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only