missing translation for 'onlineSavingsMsg'
Learn More

Silicon diOxide, purum p.a., Acid purified, Honeywell Fluka™

Catalog No. p-7072279
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002861 1 kg
6002863 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002861 Supplier Honeywell Chemicals Supplier No. 848801KG
Only null left

Purum p.a., acid purified

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 60676-86-0
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 1 kg
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08g/mol
Density 2.6g/mL at 25°C
Chemical Name or Material Silicon dioxide