missing translation for 'onlineSavingsMsg'
Learn More

Silicon Dioxide, calcined, crude, Honeywell Fluka™

Catalog No. p-7071839
Change view
Click to view available options
Quantity:
2.5 kg
50 kg
6 x 2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002038 2.5 kg
60045064 6 x 2.5 kg
6002039 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6002038 Supplier Honeywell Chemicals Supplier No. 186482.5KG
Only null left

for cleaning of platinum crucibles, calcined, crude

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 2.5 kg
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08g/mol
Chemical Name or Material Silicon Dioxide
EINECSNumber 238-878-4
RTECSNumber VV7330000