missing translation for 'onlineSavingsMsg'
Learn More

Silicon diOxide, 99%, Honeywell Fluka™

Catalog No. p-7072312
Change view
Click to view available options
Quantity:
1 kg
500 g
Catalog No. Quantity
6003166 500 g
6003165 1 kg
2 options

Catalog No. 6003166

Supplier: Honeywell Chemicals S5631500G

Only null left

∼99%, 0.5-10 μm (approx. 80% between 1-5 μm)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 500 g
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08g/mol
Percent Purity 99%
Chemical Name or Material Silicon dioxide
Hazard Statement P314
RTECSNumber VV7330000