missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Silicon dioxide

Catalog No. 3428311KG
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3428311KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3428311KG Supplier Sigma Aldrich Supplier No. 3428311KG
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicon dioxide

Specifications

Melting Point 1610°C
CAS 60676-86-0
Quantity 1 kg
Molecular Formula O2Si
Linear Formula SiO2
MDL Number MFCD00011232
Synonym Cristobalite; Quartz; Sand; Silica
Molecular Weight (g/mol) 60.08
Percent Purity 99.9% trace metals basis
Refractive Index n20/D 1.544 (literature)
Density 2.6 g/mL (at 25°C)
Show More Show Less