Learn More
Silicon carbide powder, superfine, 600 grit
CAS: 409-21-2 | CSi | 40.10 g/mol
Supplier: Thermo Scientific Chemicals A135610C
Description
Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSilicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Silicon carbide powder | |
| 409-21-2 | |
| CSi | |
| silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
| HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
| methanidylidynesilylium | |
| 9863 | |
| 40.1 | |
| 3.16 |
| superfine, 600 grit | |
| 10,000 g | |
| MFCD00049531 | |
| Insoluble in water. | |
| [C-]#[Si+] | |
| 40.10 | |
| CHEBI:29390 | |
| Odorless |
Chemical Identifiers
| 409-21-2 | |
| 40.10 | |
| HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
| 9863 | |
| [C-]#[Si+] |
| CSi | |
| MFCD00049531 | |
| silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
| CHEBI:29390 |
Safety and Handling
GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 206-991-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only