missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide, beta-phase, nanopowder, 95% min

Catalog No. p-7059063
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4388406 5 g
AA4388414 25 g
AA4388422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4388406 Supplier Thermo Scientific Chemicals Supplier No. 04388406
Only null left

CAS: 409-21-2 | CSi | 40.10 g/mol

Silicon carbide is used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Used in electronic applications, measure gas temperatures in an optical technique called thin filament pyrometry. Used in jewelry, steel production, grapheme production. It is also used in production of exponential resistors, wire sawing, construction industry, a filler in plastics and composites, ceramic sintered parts, ferrites and soft metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black to Gray
Physical Form Powder
Quantity 5 g
Merck Index 14,8492
Solubility Information Insoluble in water, alcohol; Soluble in fused alkali
IUPAC Name methanidylidynesilanylium
Formula Weight 40.1
Percent Purity ≥95%
Odor Odorless
Chemical Name or Material Silicon carbide, beta-phase
Show More Show Less
Hazard Category H315-H319-H335-H350
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 206-991-8
RTECSNumber VW0450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only