missing translation for 'onlineSavingsMsg'
Learn More

Silicic Acid, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5391470
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003919 500 g Poly Bottle
02003918 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003919 Supplier Avantor Supplier No. 032401
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
Synonym metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Quantity 500 g
Physical Form Powder
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Silicic Acid, n-Hydrate