missing translation for 'onlineSavingsMsg'
Learn More

Silica Standard, 10 ppm SiO2 (4.67 ppm Si), Ricca Chemical

Catalog No. 674032
Change view
Click to view available options
Package: Natural Poly; Size: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
674032 Package: Natural Poly; Size: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 674032 Supplier Ricca Chemical Company Supplier No. 674032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 6834-92-0, 7732-18-5
CAS Min % 98.0, 0.01
CAS Max % 100.0, 0.01
Equivalence 1 mL = 0.01 mg SiO
Molecular Formula Na2O3Si
InChI Key NTHWMYGWWRZVTN-UHFFFAOYSA-N
SMILES [O-][Si](=O)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido(oxo)silane
Molecular Weight (g/mol) 122.062
PubChem CID 23266
ChEBI CHEBI:60720
Chemical Name or Material Silica Standard
Show More Show Less