Learn More
Thermo Scientific Chemicals Silica gel, wide pore, 150 angstroms
Catalog No. AA4272618
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4272618 50 g
AA4272630 250 g
AA42726A1 1 kg
AA42726A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4272618 Supplier Thermo Scientific Chemicals Supplier No. 04272618
Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Particle Size -100/200 Mesh
Pore Size 150 Å
Packaging Bottle
pH 6.5 to 7.5
Quantity 50 g
Surface Area (Metric) 350 to 400 m2/g
Type Silica Gel
Material Silica
RUO – Research Use Only