Learn More
Silica gel, TLC high purity grade, without binder
For TLC Plate Preparation | CAS: 63231-67-4 | H2O3Si | 78.098 g/mol
Supplier: Thermo Scientific Chemicals 042756A1
Description
Silica gel, TLC high purity grade, without binder is employed for thin layer chromatography plate preparation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silica gel | |
| 63231-67-4 | |
| 1 kg | |
| MFCD00011232 | |
| Soluble in alkalies. Insoluble in water. | |
| O[Si](=O)O | |
| 78.098 | |
| TLC High Purity |
| White | |
| Powder | |
| H2O3Si | |
| metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid | |
| IJKVHSBPTUYDLN-UHFFFAOYSA-N | |
| dihydroxy(oxo)silane | |
| 14768 | |
| Odorless |
Chemical Identifiers
| 63231-67-4 | |
| 78.098 | |
| IJKVHSBPTUYDLN-UHFFFAOYSA-N | |
| 14768 | |
| O[Si](=O)O |
| H2O3Si | |
| MFCD00011232 | |
| metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid | |
| dihydroxy(oxo)silane |
Safety and Handling
H373: May cause damage to organs through prolonged or repeated exposure.
EINECSNumber : 231-545-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only