missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, TLC high purity grade, without binder

Catalog No. AA4275630
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4275630 250 g
AA42756A1 1 kg
AA42756A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4275630 Supplier Thermo Scientific Chemicals Supplier No. 04275630

For TLC Plate Preparation | CAS: 63231-67-4 | H2O3Si | 78.098 g/mol

Silica gel, TLC high purity grade, without binder is employed for thin layer chromatography plate preparation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
Synonym metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Color White
Physical Form Powder
Quantity 250 g
Solubility Information Soluble in alkalies. Insoluble in water.
Grade TLC High Purity
Odor Odorless
Chemical Name or Material Silica gel
Hazard Category H373: May cause damage to organs through prolonged or repeated exposure.
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only