missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel, Technical Grade 40, 6-14 mesh

Catalog No. 111012600
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012600 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012600 Supplier MilliporeSigma™ Supelco™ Supplier No. 2144261KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For preparative liquid chromatography

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation lresulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:a. Aquagel - pores are filled with waterb. Xerogel - by the process of evaporation, aqueous phase in the pores are removedc. Aerogel - solvent removed by supercritical extraction

Specifications

Particle Size 6/14 Mesh
Pore Size 40 Å
Grade Technical Grade 40
Packaging Poly Bottle
Quantity 1 kg
Surface Area (Metric) 500 m2/g (approx.)
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8