missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel, Spherical, 40-75 μm

Catalog No. 111012529
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012529 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012529 Supplier MilliporeSigma™ Supelco™ Supplier No. 80442100G

missing translation for 'validMainframeMsg'

High purity grade silica gel offers the lowest moisture content, tightest particle size distribution, minimal impurities and greatest lot-to-lot consistency which results in greater reproducibility of the separation process.

High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.

Specifications

Particle Size 40 to 75 μm
Grade High-purity Grade
pH 7 (5% in H2O, suspension)
Quantity 100 g
Surface Area (Metric) 250 to 350 m2/g
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8