missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel (Spherical, 100micro m) [for Column Chromatography], TCI America™

Catalog No. S08221KG
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08221KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08221KG Supplier TCI America Supplier No. S08221KG
Only null left

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Color White
Physical Form Crystal-Powder at 20°C
Material Silica Gel
Quantity 1 kg
Formula Weight 60.08
Chemical Name or Material Silica Gel (Spherical, 100micro m) [for Column Chromatography]
EINECSNumber (1)-0548
RTECSNumber VV7565000
TSCA Yes