missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica gel, preparative chromatography grade, spherical, 10 micron APS, 60 angstroms, 99.99+%
Description
Silica gel is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Spherical, S.A. 650m |
| Material | Silica Gel |
| Quantity | 5 g |
| Particle Size | 10 Micron APS, 60Å |
| Solubility Information | Insoluble in water. |
| Percent Purity | ≥99.99% |
| Grade | Preparative Chromatography |
| Surface Area | 650m2/g |
| Chemical Name or Material | Silica gel, P.V. 1.00cc/g |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H373-H319-H335 May cause damage to organs through prolonged or repeated exposure. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P260-P261-P280-P305+P351+P338-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 231-545-4 |
| RTECSNumber | VV7340000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only