missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel orange with moisture indicator free of heavy metals

Catalog No. 111012564
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012564 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012564 Supplier MilliporeSigma™ Supelco™ Supplier No. 137672.5KGR

Silica gel orange (Drying pearls Orange) is a commonly employed drying agent.

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density.

Specifications

Packaging Poly Bottle
Quantity 2.5 kg
Capacity (Metric) ≥25% absorption capacity (water)
MDL Number MFCD00011232
CAS 112926-00-8