missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Silica Gel orange with moisture indicator free of heavy metals
Description
Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density.
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Capacity (Metric) | ≥25% absorption capacity (water) |
| MDL Number | MFCD00011232 |
| CAS | 112926-00-8 |