missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel, Orange, Granular, 0.2-1 mm

Catalog No. 111012582
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012582 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012582 Supplier MilliporeSigma™ Supelco™ Supplier No. 10087500GR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For thin layer and column chromatography

Silica gel, highly porous textured matrix are generally used for synthesis of adsorbents. They are corpuscular in structure. Silica surface have a layer of hydroxyl groups and adsorbed water molecules are removed by drying at high temperature or even by treatment with suitable reagents. Silica gel surface plays significant role in catalysis, chemical separation and microelectronic fabrications.

Specifications

Particle Size 0.2 to 1 mm
Pore Size 60 Å
Grade High-purity Grade
Quantity 500 g
Surface Area (Metric) 500 m2/g (approx.)
Physical Form Granular
MDL Number MFCD00011232
CAS 112926-00-8