missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, large pore, 99.5%

Catalog No. p-7080506
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8938536 500 g
AA89385A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8938536 Supplier Thermo Scientific Chemicals Supplier No. 08938536
Only null left

CAS: 63231-67-4 | H2O3Si | 78.098 g/mol

A useful product for column chromatography (HPLC). Used as cat litter, food additive, water filtration and humidity indicator. Also a reliable substance to preserve field-collected leafs for molecular studies for variation in DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Color White
Physical Form 56 Micron,S.A. 500m
Material Silica Gel
Quantity 500 g
Particle Size 56 Micron
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Percent Purity 99.5%
Surface Area 500m2/g
Chemical Name or Material Silica gel, Large pore, P.V. 0.75cc/g
Hazard Category Warning
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-545-4
RTECSNumber VV7340000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only