missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Indicating Expose to atmosphere only when put into use., Reagents

Catalog No. 01337072
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337072 2.5 kg
01337071 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337072 Supplier Labchem Inc Supplier No. C23007002.5A5
Only null left

Silica Gel, Indicating Expose to atmosphere only when put into use., Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 112926-00-8 , 7646-79-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Solid
pH 5.5 to 9.0
Color Blue
Material Silica Gel
Quantity 2.5 kg
Packaging Poly Bottle
Specific Gravity 2.1
Chemical Name or Material Silica Gel, Indicating Expose to atmosphere only when put into use.