missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Indicating Expose to atmosphere only when put into use., Reagents

Catalog No. 01337071
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 kg
500 g
Catalog No. Quantity
01337071 500 g
01337072 2.5 kg
2 options

Catalog No. 01337071

Supplier: Labchem Inc C2300700500A5

Only null left

Silica Gel, Indicating Expose to atmosphere only when put into use., Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 112926-00-8 , 7646-79-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Solid
Color Blue
pH 5.5 to 9.0
Material Silica Gel
Quantity 500 g
Packaging Poly Bottle
Specific Gravity 2.1
Chemical Name or Material Silica Gel, Indicating Expose to atmosphere only when put into use.