missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, HPLC grade, spherical, 5 micron APS, 120 angstroms, 99.99+%

Catalog No. p-7059960
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4581006 5 g
AA4581014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4581006 Supplier Thermo Scientific Chemicals Supplier No. 04581006
Only null left

CAS: 63231-67-4 | H2O3Si | 78.098 g/mol

Silica gel is a useful product for column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Physical Form Spherical, S.A. 340m
Material Silica Gel
Quantity 5 g
Particle Size 3 Micron APS, 120Å
Solubility Information Insoluble in water.
Percent Purity ≥99.99%
Grade HPLC
Surface Area 340m2/g
Chemical Name or Material Silica gel, P.V. 1.00cc/g
Hazard Category Warning
Hazard Statement GHS H Statement
H373-H319-H335
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-545-4
RTECSNumber VV7340000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only