missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, HPLC grade, spherical, 2.2 micron APS, 80 angstroms, 99.99+%

Catalog No. p-7059955
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4579703 1 g
AA4579706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4579703 Supplier Thermo Scientific Chemicals Supplier No. 04579703
Only null left

CAS: 63231-67-4 | H2O3Si | 78.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Physical Form Spherical, S.A. 470m
Material Silica Gel
Quantity 1 g
Particle Size 2.2 Micron APS, 80Å
Percent Purity ≥99.99%
Grade HPLC
Surface Area 470m2/g
Chemical Name or Material Silica gel, P.V. 0.95cc/g
Hazard Category Warning
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-545-4
RTECSNumber VV7340000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only