missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Solstice

Catalog No. 60044822
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
6 x 1 kg
6 x 2.5 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60044822 6 x 500 g
6002004 500 g
6002002 1 kg
6002003 2.5 kg
60044820 6 x 1 kg
60044821 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60044822 Supplier Solstice Supplier No. 137676X500G

missing translation for 'validMainframeMsg'

With moisture indicator free of heavy metals

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 112926-00-8
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 6 x 500 g
UN Number NONH for all modes of transport
Chemical Name or Material Silica Gel
EINECSNumber 231-545-4