missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel, High-Purity Grade (Davisil™ Grade 710)

Catalog No. 111012607
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012607 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012607 Supplier MilliporeSigma™ Supelco™ Supplier No. 236756100G

For thin layer chromatography

High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.

Specifications

Particle Size 10.0 to 14.0 μm
Pore Size 50 to 76 Å
Grade Davisil Grade 710
Packaging Poly Bottle
pH 6.0 to 7.0 (5% in slurry)
Quantity 100 g
Surface Area (Metric) 475 to 560 m2/g
Product Line Davisil
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8
Percent Purity 99%
Show More Show Less

Davisil is a registered trademark of W.R. Grace & Co.-Conn.