missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel, High-Purity Grade (Davisil™ Grade 633), 200-425 mesh

Catalog No. 111012608
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012608 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012608 Supplier MilliporeSigma™ Supelco™ Supplier No. 236772100G

For column chromatography

High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.

Specifications

Particle Size 200/425 Mesh
Pore Size 60 Å
Grade High-purity Grade (Davisil Grade 633)
Packaging Poly Bottle
pH 6.5 (5% in slurry)
Quantity 100 g
Surface Area (Metric) 480 m2/g
Product Line Davisil
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8
Percent Purity ≥99%
Show More Show Less

Davisil is a registered trademark of W.R. Grace & Co.-Conn.