missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Silica Gel, High-Purity Grade (Davisil™ Grade 22), 60-200 mesh
Description
High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.
Specifications
Specifications
| Particle Size | 60/200 Mesh |
| Pore Size | 60 Å |
| Grade | Davisil Grade 22 |
| Packaging | Poly Bottle |
| Quantity | 1 kg |
| Surface Area (Metric) | 475 to 560 m2/g |
| Product Line | Davisil |
| Physical Form | Powder or Crystals |
| MDL Number | MFCD00011232 |
| CAS | 112926-00-8 |
Davisil is a registered trademark of W.R. Grace & Co.-Conn.