missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Silica Gel, High-Purity Grade (7754), 70-230 mesh

Catalog No. 111012572
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012572 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012572 Supplier MilliporeSigma™ Supelco™ Supplier No. 403598500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For column chromatography

High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.

Specifications

Particle Size 70/230 Mesh
Grade High-purity Grade (7754)
Packaging Poly Bottle
Quantity 500 g
Surface Area (Metric) 550 m2/g
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8