missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Silica Gel, High-Purity Grade (7749), with gypsum binder and fluorescent indicator, for thin layer chromatography
Description
High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.
Specifications
Specifications
| Pore Size | 60 Å |
| Grade | For thin layer chromatography |
| Packaging | Poly Bottle |
| Quantity | 1 kg |
| Physical Form | Powder |
| MDL Number | MFCD00011232 |
| CAS | 112926-00-8 |