missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Silica Gel, High-Purity Grade 40, 35-70 mesh
Description
High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.
Specifications
Specifications
| Particle Size | 35/70 Mesh |
| Pore Size | 90 Å |
| Technique | Chromatography |
| Grade | For column chromatography |
| Quantity | 1 kg |
| Surface Area (Metric) | 350 m2/g |
| Material | Silica |
| Physical Form | Powder or Crystals |
| MDL Number | MFCD00011232 |
| CAS | 112926-00-8 |