Learn More
Silica Gel, Grade 923, 100-200 Mesh, Spectrum™ Chemical
SIL40, 7631-86-9, SiO2
Supplier: Spectrum Chemical Mfg Cor SIL40500GMCS6
Description
Spectrum™ Chemical Silica Gel, Grade 923, 100-200 Mesh is a porous and granular form of silicon dioxide which is synthetically made from sodium silicate. In chemistry, the silica gel is used as a stationary phase in chromatography. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
| Silica Gel | |
| 1 | |
| 6 x 500 g | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| Laboratory |
| 7631-86-9 | |
| Silica Gel | |
| O2Si | |
| Amber Glass Bottle | |
| O=[Si]=O | |
| 60.08 |
Chemical Identifiers
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| silanedione |