missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel, Grade 633, 200-425 Mesh, 60A, Spectrum™ Chemical
SIL61, 7631-86-9
$376.10 - $7374.20
Chemical Identifiers
| CAS | 7631-86-9 |
|---|---|
| Molecular Formula | O2Si |
| Molecular Weight (g/mol) | 60.08 |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| IUPAC Name | silanedione |
| SMILES | O=[Si]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606204
|
Spectrum Chemical Mfg Cor
SIL61100GM |
100 g | Steel Can |
Each for $376.10
|
|
||||
|
18606206
|
Spectrum Chemical Mfg Cor
SIL615KG |
5 kg | Steel Can |
Each for $7,374.20
|
|
||||
|
18606207
|
Spectrum Chemical Mfg Cor
SIL6125KGBL |
25 kg | Steel Can |
Each for $6,462.97
|
|
||||
Description
Spectrum™ Chemical Silica Gel, Grade 633, 200-425 Mesh, 60A is a form of silicon dioxide made from sodium silicate. It is granular and porous solid and is a naturally occurring mineral. It is often used as a desiccant to control humidity and avoid spoilage. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Chemical Identifiers
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| silanedione |
Specifications
| 7631-86-9 | |
| Silica Gel | |
| O2Si | |
| Steel Can | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| Ungraded | |
| Silica Gel |
| 1 | |
| 100 g | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| 0.70-0.85 CC/G | |
| O=[Si]=O | |
| 60.08 | |
| 480 m2/g |