missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel, Grade 60, 60-200 Micron, 60A, Spectrum™ Chemical
Specifications
Specifications
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| pH | 6.0 to 8.0 |
| Quantity | 100 g |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.