missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Grade 47 (10-18 Mesh/Certified ACS), Fisher Chemical™

Catalog No. S161500
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S161500 500 g Poly Bottle
S161212 2.5 kg Poly Bottle
S16110 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S161500 Supplier Thermo Fisher Scientific Supplier No. S161500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Specifications

Chemical Name or Material Silica Gel
Color Blue
Physical Form Solid
Name Note Tel-Tale™ Desiccant, Indicating, 10-18 Mesh
CAS 7646-79-9, 7631-86-9
Material Silica Gel
Quantity 500 g
Identification Pass Test
Molecular Formula SiO2
MDL Number MFCD00011232
Mesh Size 10 to 18 Mesh
Packaging Poly Bottle
Suitability Pass Test
Grade Certified ACS
Show More Show Less