missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, functionalized, Amino-3, ca.1.4mmol/g, part.size 40-63 micron, Thermo Scientific™

Chemical Identifiers

CAS 71888-97-6
Molecular Formula C9H23NO5Si2
Molecular Weight (g/mol) 281.455
MDL Number MFCD03427643
InChI Key JIBSXWZIQTVOKJ-UHFFFAOYSA-N
PubChem CID 172930
IUPAC Name dioxosilane;3-triethoxysilylpropan-1-amine
SMILES CCO[Si](CCCN)(OCC)OCC.O=[Si]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC360180100
SDS View Documents
Thermo Scientific Chemicals
360180100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC360180250
SDS View Documents
Thermo Scientific Chemicals
360180250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC360181000
SDS View Documents
Thermo Scientific Chemicals
360181000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For solid support synthesis
Specifications

Chemical Identifiers

71888-97-6
281.455
JIBSXWZIQTVOKJ-UHFFFAOYSA-N
dioxosilane;3-triethoxysilylpropan-1-amine
C9H23NO5Si2
MFCD03427643
172930
CCO[Si](CCCN)(OCC)OCC.O=[Si]=O

Specifications

71888-97-6
C9H23NO5Si2
Glass bottle
JIBSXWZIQTVOKJ-UHFFFAOYSA-N
dioxosilane;3-triethoxysilylpropan-1-amine
172930
10g
MFCD03427643
Particle size: 40 to 63 Micron
CCO[Si](CCCN)(OCC)OCC.O=[Si]=O
281.455
Silica gel, functionalized, Amino-3
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only