missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, for drying purposes, non-toxic grade, 3-6 mm

Catalog No. p-41554
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC357400010 1 kg Plastic Bottle
AC357400050 5 kg Plastic Drum
AC357400250 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC357400010 Supplier Thermo Scientific Chemicals Supplier No. 357400010
Only null left

Silica gel, O2Si, CAS Number-7631-86-9, Silicon dioxide, 25kg, 2.0% max. (140 deg.C, 4 hrs), Adsorption capacity (relative humidity 40%): ca 20%, 620 to 800g/L, CHEBI:30563, White to Yellow, 231-545-4, 60.08, Non-toxic, VYPSYNLAJGMNEJ-UHFFFAOYSA-N, dioxosilane, 15, 8632, MFCD00011232 | CAS: 7631-86-9 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent, For column chromatography

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Silicon dioxide
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color White to Yellow
Physical Form Solid
Density 620 to 800g/L
Material Silica Gel
Quantity 1 kg
Moisture Residual moisture (140°C): < 1% (typically)
Merck Index 15, 8632
Packaging Plastic Bottle
Particle Size Particle size analysis: typically sieve residue>5mm:ca 10%, passing through 2mm: ca 1%
Solubility Information Solubility in water: insoluble.
IUPAC Name silanedione
Formula Weight 60.08
Grade Non-toxic
Water Absorption 20.0% min. (40% min. RH)
Chemical Name or Material Silica gel, 3-6mm
Show More Show Less
EINECSNumber 231-545-4
RTECSNumber EU8655000
TSCA TSCA

RUO – Research Use Only