missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical™

Catalog No. p-33390
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S162500 500 g Poly Bottle
S162212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S162500 Supplier Thermo Fisher Scientific Supplier No. S162500
Only null left

Specifications

Chemical Name or Material Silica Gel Desiccant, Indicating
Physical Form Solid
Color Blue
Name Note 4-10 Mesh, Grade 48
CAS 112926-00-8, 7646-79-9
Material Silica Gel
Quantity 500 g
Identification Pass Test
Molecular Formula SiO2
MDL Number MFCD00011232
Mesh Size 4 to 10 Mesh
Packaging Poly Bottle
Suitability Pass Test
Grade Certified ACS
Show More Show Less