missing translation for 'onlineSavingsMsg'
Learn More

Silica gel desiccant, indicating, -6+16 mesh granules

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Supplier:  Thermo Scientific Chemicals 044389A1

 View more versions of this product

Catalog No. AA44389A1

Only null left
Explore available promotions

Description

Description

Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1610°C to 1728°C
Granular
Silica Gel Desiccant
2.17 to 2.66g/cm3
1 kg
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Odorless
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Hygroscopic
60.08
CHEBI:30563
2230°C
Blue
7631-86-9
Silica Gel
O2Si
-6+16 Mesh
Soluble in alkalies. Insoluble in water.
O=[Si]=O
dioxosilane
24261
ACS

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
CHEBI:30563
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
24261
O=[Si]=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H350i-H360F

EINECSNumber : 231-545-4

RTECSNumber : VV7565000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only