missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Desiccant (60-200 Mesh/Chromatographic Grade), Fisher Chemical™

Catalog No. p-33359
Change view
Click to view available options
Quantity:
10 kg
25 kg
Packaging:
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S70410 10 kg Fiber Drum
S70425 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S70410 Supplier Thermo Fisher Scientific Supplier No. FLS70410

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Color Blue
Physical Form Solid
Material Silica Gel
Quantity 10 kg
Identification Pass Test
Mesh Size 60 to 200 Mesh
Packaging Fiber Drum
Grade Chromatography
Chemical Name or Material Silica Gel Desiccant, 60-200 Mesh
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Possible cancer hazard May cause cancer based on animal data. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1