missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Desiccant (28-200 mesh, Grade 12), Fisher Chemical

Catalog No. S157212
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S157212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S157212 Supplier Thermo Fisher Scientific Supplier No. S157 212
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Color White
Boiling Point 2230°C
Physical Form Solid
Melting Point 1710°C
pH 2.3 to 7.4
Density 2.1g/cm³
Material Silica Gel
Quantity 2.5 kg
Identification Pass Test
Mesh Size 28 to 200 Mesh
Packaging Poly Bottle
Suitability Pass Test
Chemical Name or Material Silica Gel Desiccant, 28-200 Mesh, Grade 12
Show More Show Less