missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Silica gel Grade 923 conforms to the specifications of ASTM D1319 and ASTM D2549.
Supplier: Thermo Fisher Scientific S67972X10G
Description
- Sold as a case of 3 boxes - each box contains 24 x 10 g pouches
- Suitable for use in Fluorescent Indicator Adsorption (FIA) test
- Conforms to the specifications of ASTM D1319 and ASTM D2549
- Surface area within the range of 430 - 530 m2/g
- This grade of silica gel is also available in other sizes:
- 500 g (S679500)
- 2.5 kg (S679212)
- 25 g (S67925)
Specifications
| 1710°C | |
| Solid | |
| White | |
| 100-200 Mesh | |
| 2.1g/cm³ | |
| 72 x 10 g | |
| O2Si | |
| Inclusive between 4.5% | |
| 5.0% max. | |
| O=[Si]=O | |
| 60.08 | |
| CHEBI:30563 | |
| Inclusive between 430m2/g |
| 2230°C | |
| 5.5 to 7.0 (for 5% slurry) | |
| Silica Gel Desiccant | |
| 7631-86-9 | |
| Silica Gel | |
| Pass Test | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| Pouch | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| 24261 | |
| Chromatography |
Chemical Identifiers
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| CHEBI:30563 | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| 24261 | |
| silanedione |
Safety and Handling