missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Davisil(R), Grade 635, 60-100 Mesh, 60A, Spectrum™ Chemical

SIL63, 7631-86-9

$396.13 - $10924.07

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606197
View Documents
Spectrum Chemical Mfg Cor
SIL63100GM
100 g Amber Glass Bottle
Each for $396.13
Only null left
 
18606198
View Documents
Spectrum Chemical Mfg Cor
SIL631KG
1 kg Amber Glass Bottle
Each for $2,491.87
Only null left
 
18606199
View Documents
Spectrum Chemical Mfg Cor
SIL635KG
5 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Silica Gel, Davisil, Grade 635, 60-100 Mesh, 60A , a form of silicon dioxide, is made from sodium silicate. As a granular and porous solid, it is a naturally occurring mineral and often used as a desiccant to control humidity and avoid spoilage. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silanedione

Specifications

7631-86-9
Silica Gel
O2Si
Amber Glass Bottle
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
Ungraded
1
100 g
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
60-100 Mesh
O=[Si]=O
60.08
Silica Gel, Davisil
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'