missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, for column chrom., ultra pure, 35-70 µm, 60A

Catalog No. AC360050010
Change view
Click to view available options
Quantity:
1 kg
5 kg
30 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC360050010 1 kg
AC360050050 5 kg
AC360050300 30 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC360050010 Supplier Thermo Scientific Chemicals Supplier No. 360050010

For column chromatography. | CAS: 7631-86-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7631-86-9

Specifications

Particle Size 40 to 60 μm
Pore Size 60 Å
Packaging Plastic Bottle
Quantity 1 kg
Type Silica Gel
Material Silica Gel

RUO – Research Use Only