missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, for column chromatography,ultra pure, 40-63μm, 40A

Numéro de catalogue. p-3756252
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC360035000 500 g
AC360031000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC360035000 Fournisseur Thermo Scientific Chemicals Code fournisseur 360035000
Il en reste null

CAS: 7631-86-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

Identifiants chimiques

CAS 7631-86-9

Spécifications

Particle Size 40 to 60 μm
Pore Size 40 Å
Packaging Plastic Bottle
Quantity 500 g
Type Silica Gel
Material Silica Gel
For Use With (Application) Column Chromatography
Molecular Formula O2Si
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 60.08
Chemical Name or Material Silica gel
Afficher plus de résultats Afficher moins de résultats

RUO – Research Use Only